Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC700564
Entrez Gene ID
700564
Gene Name
melanoma antigen preferentially expressed in tumors
Gene Aliases
EGK_02857, PRAME
Chromosome Location
Chr.10: 30108280 - 30120925 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001091109.4 XP_001091109.1 - 621 144
GenBank mRNA JV044986.1 - - 134 144
JV647005.1 - - 144 144

View Full Product Details