Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP50
Entrez Gene ID
713282
Gene Name
nucleoporin 50
Gene Aliases
-
Chromosome Location
Chr.10: 6323796 - 6348857 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265689.2 NP_001252618.1 - 1253 98
XM_015150553.2 XP_015006039.1 - 1145 98
GenBank mRNA JU332594.1 - - 1368 98
JU474062.1 - - 1392 98
JV045976.1 - - 1266 98
JV667328.1 - - 1253 98

View Full Product Details