Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPCS1
Entrez Gene ID
696426
Gene Name
signal peptidase complex subunit 1
Gene Aliases
-
Chromosome Location
Chr.2: 108873778 - 108876170 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193749.1 NP_001180678.1 - 432 149
GenBank mRNA JU334140.1 - - 269 149
JV046837.1 - - 285 149
JV579453.1 - - 301 149
JV649239.1 - - 311 149

View Full Product Details