Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSUH2
Entrez Gene ID
702279
Gene Name
ssu-2 homolog
Gene Aliases
C2H3orf32
Chromosome Location
Chr.2: 140003508 - 140029180 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001097726.4 XP_001097726.3 - 1438 143

View Full Product Details