Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASIC5
Entrez Gene ID
699617
Gene Name
acid sensing ion channel subunit family member 5
Gene Aliases
ACCN5
Chromosome Location
Chr.5: 153448185 - 153484767 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092816.2 XP_001092816.2 - 1027 81

View Full Product Details