Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP22
Entrez Gene ID
705824
Gene Name
dual specificity phosphatase 22
Gene Aliases
-
Chromosome Location
Chr.4: 169746824 - 169815601 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260542.1 NP_001247471.1 - 130 73
XM_015135394.2 XP_014990880.1 - 282 73
XM_028846751.1 XP_028702584.1 - 185 73
GenBank mRNA JV044251.1 - - 130 73
JV635561.1 - - 474 73

View Full Product Details