Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB22A
Entrez Gene ID
699283
Gene Name
RAB22A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.10: 93475814 - 93536483 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU322247.1 - - 720 132
JU476528.1 - - 717 132

View Full Product Details