Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NGB
Entrez Gene ID
641331
Gene Name
neuroglobin
Gene Aliases
-
Chromosome Location
Chr.7: 139373359 - 139377656 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035514.1 NP_001030591.1 - 313 75
GenBank mRNA JU322051.1 - - 417 75
JV530004.1 - - 321 75

View Full Product Details