Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENPP5
Entrez Gene ID
706238
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase family member 5
Gene Aliases
EGK_14969
Chromosome Location
Chr.4: 123210786 - 123219902 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015136489.2 XP_014991975.1 - 1068 162
XM_015136490.2 XP_014991976.1 - 1122 162
XM_015136491.2 XP_014991977.1 - 942 162
XM_015136492.2 XP_014991978.1 - 923 162
GenBank mRNA JU323211.1 - - 1022 162
JU472790.1 - - 865 162
JV601365.1 - - 1022 162

View Full Product Details