Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGM5
Entrez Gene ID
698589
Gene Name
phosphoglucomutase 5
Gene Aliases
-
Chromosome Location
Chr.15: 83787693 - 83967342 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028835225.1 XP_028691058.1 - 404 115

View Full Product Details