Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS11
Entrez Gene ID
697743
Gene Name
mitochondrial ribosomal protein S11
Gene Aliases
-
Chromosome Location
Chr.7: 67242382 - 67253260 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261578.2 NP_001248507.1 - 490 122
XM_015142938.2 XP_014998424.1 - 682 122
XM_015142939.2 XP_014998425.1 - 1620 122
GenBank mRNA JU335969.1 - - 486 122
JU476852.1 - - 490 122
JV659122.1 - - 490 122

View Full Product Details