Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CERS4
Entrez Gene ID
706728
Gene Name
ceramide synthase 4
Gene Aliases
LASS4
Chromosome Location
Chr.19: 8112124 - 8162328 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015122718.2 XP_014978204.1 - 1062 137
XM_015122719.2 XP_014978205.1 - 937 137
XM_015122720.2 XP_014978206.1 - 1093 137
GenBank mRNA JU336384.1 - - 780 137
JV048252.1 - - 746 137
JV696363.1 - - 748 137

View Full Product Details