Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF20
Entrez Gene ID
708719
Gene Name
transmembrane 4 L six family member 20
Gene Aliases
-
Chromosome Location
Chr.12: 114808988 - 114825219 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194504.2 NP_001181433.1 - 275 115

View Full Product Details