Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRR5L
Entrez Gene ID
716934
Gene Name
proline rich 5 like
Gene Aliases
-
Chromosome Location
Chr.14: 28993084 - 29170582 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194662.2 NP_001181591.1 - 576 146
XM_015114477.2 XP_014969963.1 - 2914 146
XM_015114478.2 XP_014969964.1 - 938 146
XM_015114480.2 XP_014969966.1 - 1999 146
XM_015114481.2 XP_014969967.1 - 727 146
XM_028832805.1 XP_028688638.1 - 938 146
GenBank mRNA JU321244.1 - - 628 146
JV723732.1 - - 448 146

View Full Product Details