Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRA10AC1
Entrez Gene ID
700389
Gene Name
FRA10A associated CGG repeat 1
Gene Aliases
C10orf4, C9H10orf4
Chromosome Location
Chr.9: 93829024 - 93863246 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015147634.2 XP_015003120.1 - 1021 137
XM_015147636.2 XP_015003122.1 - 1320 137
XM_028853355.1 XP_028709188.1 - 930 137
XM_028853356.1 XP_028709189.1 - 907 137
GenBank mRNA JU322797.1 - - 1073 137

View Full Product Details