Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP21
Entrez Gene ID
695797
Gene Name
matrix metallopeptidase 21
Gene Aliases
MMP23A
Chromosome Location
Chr.9: 126200772 - 126210090 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193756.2 NP_001180685.2 - 694 94

View Full Product Details