Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XXYLT1
Entrez Gene ID
708627
Gene Name
xyloside xylosyltransferase 1
Gene Aliases
C3orf21
Chromosome Location
Chr.2: 2694254 - 2888999 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194076.3 NP_001181005.1 - 608 139
GenBank mRNA JU327309.1 - - 144 139
JV643405.1 - - 229 139

View Full Product Details