Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H9orf43
Entrez Gene ID
704968
Gene Name
chromosome 15 C9orf43 homolog
Gene Aliases
C9orf43
Chromosome Location
Chr.15: 29411991 - 29431291 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001101033.4 XP_001101033.1 - 1430 95
XM_015117039.2 XP_014972525.1 - 1277 95
XM_028834963.1 XP_028690796.1 - 2011 95
GenBank mRNA JU483931.1 - - 80 95

View Full Product Details