Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACC1
Entrez Gene ID
700834
Gene Name
laccase domain containing 1
Gene Aliases
C13orf31, C17H13orf31
Chromosome Location
Chr.17: 22451589 - 22591683 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193989.2 NP_001180918.1 - 584 149
XM_015120901.2 XP_014976387.1 - 668 149
XM_028837113.1 XP_028692946.1 - 660 149
GenBank mRNA JU323204.1 - - 569 149

View Full Product Details