Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MROH2B
Entrez Gene ID
697286
Gene Name
maestro heat like repeat family member 2B
Gene Aliases
HEATR7B2
Chromosome Location
Chr.6: 40877565 - 40952656 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139951.2 XP_014995437.2 - 3363 70

View Full Product Details