Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA12
Entrez Gene ID
701708
Gene Name
serpin family A member 12
Gene Aliases
-
Chromosome Location
Chr.7: 156661555 - 156678236 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001099453.4 XP_001099453.1 - 1113 116

View Full Product Details