Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFB6
Entrez Gene ID
710835
Gene Name
BPI fold containing family B member 6
Gene Aliases
BPIL3, EGK_02510
Chromosome Location
Chr.10: 68272679 - 68284724 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028828765.1 XP_028684598.1 - 194 71

View Full Product Details