Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPTL5
Entrez Gene ID
702036
Gene Name
angiopoietin like 5
Gene Aliases
-
Chromosome Location
Chr.14: 95168986 - 95186568 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115536.2 XP_014971022.2 - 634 148

View Full Product Details