Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRG1L
Entrez Gene ID
114677490
Gene Name
tumor protein p63 regulated 1 like
Gene Aliases
-
Chromosome Location
Chr.1: 220889158 - 220894619 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028847129.1 XP_028702962.1 - 291 74

Gene Symbol
TPRG1L
Entrez Gene ID
695045
Gene Name
tumor protein p63 regulated 1 like
Gene Aliases
-
Chromosome Location
Chr.1: 30447 - 35361 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260515.1 NP_001247444.1 - 345 74
XM_015134959.2 XP_014990445.1 - 265 74
GenBank mRNA JV045147.1 - - 345 74
JV045148.1 - - 345 74
JV045149.1 - - 345 74
JV602229.1 - - 356 74

View Full Product Details