Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA5
Entrez Gene ID
708435
Gene Name
sperm acrosome associated 5
Gene Aliases
EGK_20455, LYZE, PNPK6288
Chromosome Location
Chr.X: 47946818 - 47952303 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015127305.2 XP_014982791.1 - 507 96
XM_028842172.1 XP_028698005.1 - 501 96
GenBank mRNA JU323909.1 - - 391 96

View Full Product Details