Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD14
Entrez Gene ID
697482
Gene Name
CD14 molecule
Gene Aliases
-
Chromosome Location
Chr.6: 138012823 - 138014253 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130433.2 NP_001123905.1 - 92 72
GenBank mRNA CO583835.1 - - 96 72

View Full Product Details