Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINGO4
Entrez Gene ID
711472
Gene Name
leucine rich repeat and Ig domain containing 4
Gene Aliases
EGK_01271
Chromosome Location
Chr.1: 99285029 - 99291888 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001108832.4 XP_001108832.2 - 231 81
GenBank mRNA JU485676.1 - - 64 81

View Full Product Details