Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4F3
Entrez Gene ID
696525
Gene Name
olfactory receptor, family 4, subfamily F, member 3
Gene Aliases
-
Chromosome Location
Chr.7: 81294618 - 81295605 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193784.1 NP_001180713.1 - 353 144

View Full Product Details