Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEUROD1
Entrez Gene ID
704954
Gene Name
neuronal differentiation 1
Gene Aliases
EGK_04592
Chromosome Location
Chr.12: 68767705 - 68772428 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001101116.4 XP_001101116.1 - 754 81
GenBank mRNA JV469622.1 - - 130 81

View Full Product Details