Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SYNGR3
Entrez Gene ID
694003
Gene Name
synaptogyrin 3
Gene Aliases
-
Chromosome Location
Chr.20: 2014979 - 2018481 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204764.1 NP_001191693.1 - 226 86
GenBank mRNA BQ807465.1 - - 226 86
JV667986.1 - - 216 86

View Full Product Details