Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXHXorf48
Entrez Gene ID
709338
Gene Name
chromosome X open reading frame 48
Gene Aliases
CXorf48
Chromosome Location
Chr.X: 131544821 - 131557287 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194153.1 NP_001181082.1 - 650 142

View Full Product Details