Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC31A1
Entrez Gene ID
706058
Gene Name
solute carrier family 31 member 1
Gene Aliases
EGK_07412
Chromosome Location
Chr.15: 29577435 - 29615211 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257506.1 NP_001244435.1 - 304 143
GenBank mRNA JU332952.1 - - 304 143
JU332953.1 - - 304 143
JU473239.1 - - 281 143
JV046212.1 - - 316 143
JV642423.1 - - 305 143

View Full Product Details