Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNAQ
Entrez Gene ID
705680
Gene Name
G protein subunit alpha q
Gene Aliases
-
Chromosome Location
Chr.15: 93081310 - 93404683 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261595.1 NP_001248524.1 - 1472 170
XM_028834293.1 XP_028690126.1 - 1344 173
GenBank mRNA JU334059.1 - - 1468 173
JU475226.1 - - 1472 170
JV680812.1 - - 1467 170

View Full Product Details