Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD151
Entrez Gene ID
574139
Gene Name
CD151 molecule
Gene Aliases
PETA-3
Chromosome Location
Chr.14: 887192 - 892869 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001361174.2 NP_001348103.1 - 433 71
NM_001361175.2 NP_001348104.1 - 429 71
NM_001361176.2 NP_001348105.1 - 491 71
NM_001361177.1 NP_001348106.1 - 496 71
XM_015113287.2 XP_014968773.1 - 587 71
XM_015113289.1 XP_014968775.1 - 479 71
XM_028832488.1 XP_028688321.1 - 578 71
XM_028832489.1 XP_028688322.1 - 597 71
GenBank mRNA AF275665.1 - - 381 71
JU471378.1 - - 431 71
JV048827.1 - - 379 71
JV634758.1 - - 358 71
JV638340.1 - - 443 71
JV669377.1 - - 433 71

View Full Product Details