Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM15
Entrez Gene ID
712497
Gene Name
tripartite motif containing 15
Gene Aliases
-
Chromosome Location
Chr.4: 139725926 - 139734557 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114948.1 NP_001108420.1 - 471 99

View Full Product Details