Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLR10
Entrez Gene ID
697980
Gene Name
toll like receptor 10
Gene Aliases
EGK_15647
Chromosome Location
Chr.5: 37910552 - 37921489 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130434.1 NP_001123906.1 - 2344 81
XM_015138153.2 XP_014993639.2 - 2619 81
GenBank mRNA AB445686.1 - - 2344 81

View Full Product Details