Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR7E24
Entrez Gene ID
709658
Gene Name
olfactory receptor, family 7, subfamily E, member 24
Gene Aliases
-
Chromosome Location
Chr.19: 9069472 - 9070490 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194036.3 NP_001180965.3 - 54 79

View Full Product Details