Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNPY1
Entrez Gene ID
718795
Gene Name
canopy FGF signaling regulator 1
Gene Aliases
EGK_14381
Chromosome Location
Chr.3: 181500092 - 181536824 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193382.1 NP_001180311.1 - 146 171
XM_015135355.2 XP_014990841.1 - 374 171

View Full Product Details