Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP12-4
Entrez Gene ID
712284
Gene Name
keratin associated protein 12-4
Gene Aliases
-
Chromosome Location
Chr.3: 2069551 - 2069889 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194238.1 NP_001181167.1 - 150 70

View Full Product Details