Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QPCTL
Entrez Gene ID
715134
Gene Name
glutaminyl-peptide cyclotransferase like
Gene Aliases
-
Chromosome Location
Chr.19: 45759956 - 45774454 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194501.2 NP_001181430.1 - 1542 67
XM_015124124.2 XP_014979610.1 - 1537 67
XM_015124126.2 XP_014979612.1 - 817 67

View Full Product Details