Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK9
Entrez Gene ID
100424925
Gene Name
kallikrein related peptidase 9
Gene Aliases
-
Chromosome Location
Chr.19: 50893106 - 50899307 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194833.1 NP_001181762.1 - 46 61

View Full Product Details