Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCAND1
Entrez Gene ID
711046
Gene Name
SCAN domain containing 1
Gene Aliases
EGK_02452
Chromosome Location
Chr.10: 71195567 - 71196425 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190908.2 NP_001177837.2 - 39 61

View Full Product Details