Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSP90AB1
Entrez Gene ID
702293
Gene Name
heat shock protein 90 alpha family class B member 1
Gene Aliases
-
Chromosome Location
Chr.4: 125076090 - 125084774 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195533.1 NP_001182462.1 - 101 81
GenBank mRNA CB230592.1 - - 104 81
JV504958.1 - - 93 81
JV667161.1 - - 93 81

View Full Product Details