Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR38
Entrez Gene ID
693858
Gene Name
WD repeat domain 38
Gene Aliases
-
Chromosome Location
Chr.15: 15816541 - 15821565 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193696.3 NP_001180625.2 - 134 86

View Full Product Details