Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC189
Entrez Gene ID
711827
Gene Name
coiled-coil domain containing 189
Gene Aliases
C16orf93, C20H16orf93
Chromosome Location
Chr.20: 28168707 - 28172921 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199128.1 NP_001186057.1 - 537 67

View Full Product Details