Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CD1A
- Entrez Gene ID
-
698438
- Gene Name
- CD1a molecule
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 92753887 - 92770766 on Build Mmul_10
- Gene Symbol
- LOC709286
- Entrez Gene ID
-
709286
- Gene Name
- T-cell surface glycoprotein CD1a
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 92684903 - 92689442 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU496031.1 |
- |
- |
108 |
92 |
View Full Product Details