Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSGIN2
Entrez Gene ID
695376
Gene Name
oxidative stress induced growth inhibitor family member 2
Gene Aliases
-
Chromosome Location
Chr.8: 90295712 - 90320885 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260770.1 NP_001247699.1 - 234 85
XM_015145630.2 XP_015001116.1 - 721 85
XM_015145631.2 XP_015001117.1 - 407 85
XM_015145632.2 XP_015001118.1 - 259 85
GenBank mRNA JU336215.1 - - 241 85
JU336216.1 - - 241 85
JU477057.1 - - 218 85
JV048147.1 - - 234 85
JV660467.1 - - 422 85

View Full Product Details