Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2E1
Entrez Gene ID
713562
Gene Name
general transcription factor IIE subunit 1
Gene Aliases
-
Chromosome Location
Chr.2: 155503423 - 155546884 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260889.1 NP_001247818.1 - 728 104
XM_028843353.1 XP_028699186.1 - 742 104
GenBank mRNA JU321059.1 - - 747 104
JU473756.1 - - 729 104
JU473757.1 - - 729 104
JV043413.1 - - 728 104

View Full Product Details