Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- COMT
- Entrez Gene ID
-
712548
- Gene Name
- catechol-O-methyltransferase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 32673713 - 32702017 on Build Mmul_10
- Gene Symbol
- ARVCF
- Entrez Gene ID
-
712399
- Gene Name
- ARVCF delta catenin family member
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 32618496 - 32673815 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU325911.1 |
- |
- |
5052 |
61 |
JV582160.1 |
- |
- |
4162 |
61 |
View Full Product Details