Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM7
Entrez Gene ID
695401
Gene Name
RNA binding motif protein 7
Gene Aliases
EGK_06917
Chromosome Location
Chr.14: 107552617 - 107561974 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261071.1 NP_001248000.1 - 359 83
XM_015115705.2 XP_014971191.1 - 383 83
GenBank mRNA JU332435.1 - - 359 83
JU473900.1 - - 359 83

View Full Product Details